Ifenprodil hemitartrate, 99%, Thermo Scientific Chemicals
Ifenprodil hemitartrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ifenprodil hemitartrate, 99%, Thermo Scientific Chemicals

NMDA receptor antagonist, acting at the polyamine site
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Catalog number J60932.MA
also known as J60932-MA
Price (USD)/ Each
97.10
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
97.10
Add to cart
Ifenprodil hemitartrate, 99%, Thermo Scientific Chemicals
Catalog numberJ60932.MA
Price (USD)/ Each
97.10
-
Add to cart
Specifications
Chemical Name or MaterialIfenprodil hemitartrate
CAS23210-56-2
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
NMDA receptor antagonist, acting at the polyamine site

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only