Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)
Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)
Thermo Scientific Chemicals

Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
25 mL
Catalog number J60933.AD
also known as J60933-AD
Price (USD)/ Each
107.00
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
107.00
Add to cart
Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)
Catalog numberJ60933.AD
Price (USD)/ Each
107.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAgarose gel loading dye
Name Noteglycerol based, Xylene Cyanol FF free
Physical FormLiquid
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
View more
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

Solubility
Fully misicible in water.

Notes
No Xylene cyanol FF. Contains 0.25% bromophenol blue, 30% glycerol.
RUO – Research Use Only