Thermo Scientific Chemicals

SB 202190, 99+%, Thermo Scientific Chemicals

Catalog number: J60950.MF
50 mg, Each
Thermo Scientific Chemicals

SB 202190, 99+%, Thermo Scientific Chemicals

Catalog number: J60950.MF
50 mg, Each
Quantity
Catalog number: J60950.MF
also known as J60950-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
152121-30-7
IUPAC Name
4-[4-(4-fluorophenyl)-5-(pyridin-4-yl)-2,3-dihydro-1H-imidazol-2-ylidene]cyclohexa-2,5-dien-1-one
Molecular Formula
C20H14FN3O
InChI Key
NJNKPVPFGLGHPA-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)C1=C(NC(N1)=C1C=CC(=O)C=C1)C1=CC=NC=C1
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
≥99.0%
Form
Crystalline powder or powder or granules
Proton NMR
Conforms to structure

Description

Inhibits p38 kinase activity through competition with ATP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text