Thermo Scientific Chemicals

Acetate Buffer (1 M, pH 5.0)

Catalog number: J60964.AK
250mL, Each
Thermo Scientific Chemicals

Acetate Buffer (1 M, pH 5.0)

Catalog number: J60964.AK
250mL, Each
Quantity
Catalog number: J60964.AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-09-3
IUPAC Name
sodium acetate
Molecular Formula
C2H3NaO2
InChI Key
VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES
[Na+].CC([O-])=O
Form
Clear Liquid
Composition
Acetic Acid/Sodium Acetate: 1 M
Comment
Acetate buffer is made by dissolving ingredient in18.2 megahom, UV treated water, followed by submicron filtration
pH
4.8-5.2

Description

For precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Acetate buffers for amine coupling of proteins to Biacore sensor chip surfaces. Studies on microbial pili suggest that sodium acetate suppressed K99 production in E. coli strains cultured on a minimal medium. Experiments have shown that when added to food waste composting systems, NaOAc may be effective at counteracting the adverse effects of organic acids produced in the composting process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Acetate buffers for amine coupling of proteins to Biacore sensor chip surfaces. Studies on microbial pili suggest that sodium acetate suppressed K99 production in E. coli strains cultured on a minimal medium. Experiments have shown that when added to food waste composting systems, NaOAc may be effective at counteracting the adverse effects of organic acids produced in the composting process.

Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text