Thermo Scientific Chemicals

(S)-2-(1-Hydroxyethyl)pyridine, Thermo Scientific Chemicals

Catalog number: J60980.03
1 g, Each
Thermo Scientific Chemicals

(S)-2-(1-Hydroxyethyl)pyridine, Thermo Scientific Chemicals

Catalog number: J60980.03
1 g, Each
Quantity
Catalog number: J60980.03
also known as J60980-03
Price (USD)
Quantity
-
Have Questions?

Description

Substituted 2-Pyridinemethanol derivatives are the potent and selective phosphodiesterase-4 inhibitors. Reaction of hydrous nitrosylruthenium trichloride with 2-pyridinemethanol gave ruthenium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted 2-Pyridinemethanol derivatives are the potent and selective phosphodiesterase-4 inhibitors. Reaction of hydrous nitrosylruthenium trichloride with 2-pyridinemethanol gave ruthenium complexes.

Solubility
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text