n-Octyl-beta-D-thioglucopyranoside, 98+%, Thermo Scientific Chemicals
n-Octyl-beta-D-thioglucopyranoside, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Octyl-beta-D-thioglucopyranoside, 98+%, Thermo Scientific Chemicals

Non-ionic detergent used for the solubilization of membrane proteins
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
2.5 g
10 g
Catalog number J61028.06
also known as J61028-06
Price (USD)/ Each
294.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
294.00
Add to cart
n-Octyl-beta-D-thioglucopyranoside, 98+%, Thermo Scientific Chemicals
Catalog numberJ61028.06
Price (USD)/ Each
294.00
-
Add to cart
Specifications
Chemical Name or Materialn-Octyl-beta-D-thioglucopyranoside
CAS85618-21-9
Melting Point128°C
ColorWhite
Recommended StorageAmbient temperatures
View more
n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

Solubility
Soluble in ethanol, water or methanol

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. K.H.Weisgraber; Y.M.Newhouse; A.McPherson. Crystallization and preliminary X-ray analysis of human plasma apolipoprotein C-I. Journal of Molecular Biology. 1994, 236,(1), 382-384.
  2. Sanghamitra Mukhopadhyay; David Good; Richard D.Miller; James E.Graham; Sarah A.Mathews; Peter Timms; James T.Summersgill. Identification of Chlamydia pneumoniae proteins in the transition from reticulate to elementary body formation. Molecular & Cellular Proteomics. 2006, 5,(12), 2311-2318.