Thermo Scientific Chemicals

Acetate Buffer (1 M, pH 5.5)

Catalog number: J61033.AK
250mL, Each
Thermo Scientific Chemicals

Acetate Buffer (1 M, pH 5.5)

Catalog number: J61033.AK
250mL, Each
Quantity
Catalog number: J61033.AK
also known as J61033-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-09-3
IUPAC Name
sodium acetate
Molecular Formula
C2H3NaO2
InChI Key
VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES
[Na+].CC([O-])=O
Appearance (Color)
Clear colorless
Form
Liquid
Concentration
1M
pH
5.35-5.65

Description

Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.

Notes
Contains sodium acetate and glacial acetic acid, pH 5.5
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text