Thermo Scientific Chemicals

1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals

Catalog number: J61034.AH
200 mL, Each
Thermo Scientific Chemicals

1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals

Catalog number: J61034.AH
200 mL, Each
Quantity
Catalog number: J61034.AH
also known as J61034-AH
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
288-94-8
IUPAC Name
2H-1,2,3,4-tetrazole
Molecular Formula
CH2N4
InChI Key
KJUGUADJHNHALS-UHFFFAOYSA-N
SMILES
N1N=CN=N1
Appearance (Color)
Clear colorless
Form
Liquid
Water Content (Karl Fischer Titration)
≤ 60 ppm
Comment
Filtered under argon through a 0.2micron filter
Concentration
0.45 - 0.50M tetrazole in acetonitrile

Description

1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

Solubility
Miscible with water.

Notes
Incompatible with reducing agents and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text