Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Alkaline Phosphatase, calf intestine, EIA Grade

Catalog number: J61037.LB0
5mg, Each
Thermo Scientific Chemicals

Alkaline Phosphatase, calf intestine, EIA Grade

Catalog number: J61037.LB0
5mg, Each
Quantity
Catalog number: J61037.LB0
also known as J61037-#0
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9001-78-9
IUPAC Name
2-{[1-(2-{[1-(2-aminopropanoyl)pyrrolidin-2-yl]formamido}acetyl)pyrrolidin-2-yl]formamido}-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Formula
C21H36N8O6
InChI Key
ITZMJCSORYKOSI-UHFFFAOYNA-N
SMILES
CC(N)C(=O)N1CCCC1C(=O)NCC(=O)N1CCCC1C(=O)NC(CCCN=C(N)N)C(O)=O
RNase
None detected
units/mgP
not less than 3,000 units/mg protein
DNase
None detected

Description

Alkaline Phosphatase, calf intestine, EIA GradeDephosphorylation of cloning vector DNA to prevent recircularization during ligation, dephosphorylation of DNA prior to end-labeling using T4 Polynucleotide Kinase, treatment of dNTPs in PCR reactions prior to sequencing or SNP analysis, dephosphorylation of DNA and RNA. It is also used to remove the 5?-terminal phosphate from nucleic acids during molecular cloning reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text