Thioflavine T, tech. 75%, Thermo Scientific Chemicals
Thioflavine T, tech. 75%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thioflavine T, tech. 75%, Thermo Scientific Chemicals

A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J61043.06
also known as J61043-06
Price (USD)/ Each
25.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
25.50
Add to cart
Thioflavine T, tech. 75%, Thermo Scientific Chemicals
Catalog numberJ61043.06
Price (USD)/ Each
25.50
-
Add to cart
Chemical Identifiers
CAS2390-54-7
IUPAC Name2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
Molecular FormulaC17H19ClN2S
InChI KeyJADVWWSKYZXRGX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Lambda max in water: 409-415 nm
Solution TestSolubility: Conforms
FormPowder
Appearance (Color)Yellow to orange to brown
Identification (FTIR)Conforms
A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only