Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals

Useful as a cryoprotectant. Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number J61059.K2
also known as J61059-K2
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
Add to cart
Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Catalog numberJ61059.K2
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥99.0%
DNaseNone detected
Impurity contentChloride, Cl: ≤10ppm
RNaseNone detected
Appearance (Color)Clear colorless
View more
Useful as a cryoprotectant.Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component. Further, it is used as a solvent, emollient, pharmaceutical agent and sweetening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only