Thermo Scientific Chemicals

Pyronin Y, Thermo Scientific Chemicals

Catalog number: J61068.09
10 g, Each
Thermo Scientific Chemicals

Pyronin Y, Thermo Scientific Chemicals

Catalog number: J61068.09
10 g, Each
Quantity
Catalog number: J61068.09
also known as J61068-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92-32-0
IUPAC Name
3,6-bis(dimethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular Formula
C17H19ClN2O
InChI Key
INCIMLINXXICKS-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)C1=CC2=[O+]C3=CC(=CC=C3C=C2C=C1)N(C)C
Appearance (Color)
Green to dark green or red
lambda-max (UV-Vis)
546-551nm
Specific Absorbance (E 1%/1cm)
≥48000 (Lambda max)
Form
Powder

Description

Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.

Solubility
Soluble in water. Also soluble in methyl cellosolve (9 mg/mL) and ethanol (3 mg/mL).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text