Thermo Scientific Chemicals

Tetranitro blue tetrazolium chloride, 97%

Catalog number: J61080.MC
100 mg, Each
Thermo Scientific Chemicals

Tetranitro blue tetrazolium chloride, 97%

Catalog number: J61080.MC
100 mg, Each
Quantity
Catalog number: J61080.MC
also known as J61080-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1184-43-6
IUPAC Name
2-{4'-[3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Molecular Formula
C40H28Cl2N12O10
InChI Key
VCESGVLABVSDRO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O
Purity
≥96.0%
Appearance (Color)
Yellow
Infrared spectrum
Conforms
Form
Powder or crystals or crystalline powder

Description

Redox indicator for enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text