Thermo Scientific Chemicals

Histamine diphosphate, 98%, Thermo Scientific Chemicals

Catalog number: J61088.06
5 g, Each
Thermo Scientific Chemicals

Histamine diphosphate, 98%, Thermo Scientific Chemicals

Catalog number: J61088.06
5 g, Each
Quantity
Catalog number: J61088.06
also known as J61088-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-74-1
Appearance (Color)
White to off-white
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥97.5%

Description

Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text