Thermo Scientific Chemicals

Adenosine-5'-triphosphate disodium salt hydrate, 98%

Catalog number: J61125.06
5 g, Each
Thermo Scientific Chemicals

Adenosine-5'-triphosphate disodium salt hydrate, 98%

Catalog number: J61125.06
5 g, Each
Quantity
Catalog number: J61125.06
also known as J61125-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34369-07-8
IUPAC Name
disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C10H14N5Na2O13P3
InChI Key
TTWYZDPBDWHJOR-WCYUCLFNNA-L
SMILES
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Appearance (Color)
White to cream
Form
Powder or crystalline powder
Assay from Supplier's CofA
≥98.0% (HPLC)
Identification (FTIR)
Conforms to structure
Water Content (Karl Fischer Titration)
≤10%

Description

Adenosine-5'-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

Solubility
Soluble in water (110 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), DMF (sparingly), and PBS pH 7.2 (∼10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text