Thermo Scientific Chemicals

TRIS, 0.5M buffer soln., pH 8.8

Catalog number: J61144.K2
1 L, Each
Thermo Scientific Chemicals

TRIS, 0.5M buffer soln., pH 8.8

Catalog number: J61144.K2
1 L, Each
Quantity
Catalog number: J61144.K2
also known as J61144-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-86-1
IUPAC Name
2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C4H11NO3
InChI Key
LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES
NC(CO)(CO)CO
pH
8.8 ± 0.2
Form
Clear liquid
Comment
Filtered through 0.22 micron filter

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text