Sodium phosphate, 0.5M buffer soln., pH 8.5
Sodium phosphate, 0.5M buffer soln., pH 8.5
Thermo Scientific Chemicals

Sodium phosphate, 0.5M buffer soln., pH 8.5

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number J61151.AP
also known as J61151-AP
Price (USD)/ Each
173.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
173.00
Add to cart
Sodium phosphate, 0.5M buffer soln., pH 8.5
Catalog numberJ61151.AP
Price (USD)/ Each
173.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialSodium phosphate buffer soln.
Recommended StorageAmbient temperatures
Concentration0.5 M
TSCAYes
pH8.5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only