Met-Enkephalin
Thermo Scientific Chemicals

Met-Enkephalin

Have Questions?
Change viewbuttonViewtableView
Quantity:
50mg
25mg
125mg
Catalog number J61154.MF
also known as J61154-MF
Price (USD)/ Each
314.00
-
Add to cart
Quantity:
50mg
Request bulk or custom format
Price (USD)/ Each
314.00
Add to cart
Met-Enkephalin
Catalog numberJ61154.MF
Price (USD)/ Each
314.00
-
Add to cart
Specifications
CAS58569-55-4
ColorWhite
FormatPowder
Molecular Weight (g/mol)573.665
Product TypeMet-Enkephalin
View more
Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors
RUO – Research Use Only
Contents & Storage
Store at -20°C