Thermo Scientific Chemicals

Brucine, Thermo Scientific Chemicals

Catalog number: J61178.22
100 g, Each
Thermo Scientific Chemicals

Brucine, Thermo Scientific Chemicals

Catalog number: J61178.22
100 g, Each
Quantity
Catalog number: J61178.22
also known as J61178-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
357-57-3
IUPAC Name
(1R,18S,20R,21R)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one
Molecular Formula
C23H26N2O4
InChI Key
RRKTZKIUPZVBMF-PLNGPGDESA-N
SMILES
COC1=CC2=C(C=C1OC)[C@@]13CCN4CC5=CCOC6CC(=O)N2C1[C@H]6[C@H]5C[C@@H]34
Sulfated Ash
1.0% max.
Assay (unspecified)
98.0% min.

Description

Brucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.

Solubility
Soluble in alcohol, chloroform, and benzene, slightly soluble in water, ether, and glycerol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text