Thermo Scientific Chemicals

Benzydamine hydrochloride, Thermo Scientific Chemicals

Catalog number: J61180.06
5 g, Each
Thermo Scientific Chemicals

Benzydamine hydrochloride, Thermo Scientific Chemicals

Catalog number: J61180.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61180.06
also known as J61180-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-69-4
IUPAC Name
hydrogen {3-[(1-benzyl-1H-indazol-3-yl)oxy]propyl}dimethylamine chloride
Molecular Formula
C19H24ClN3O
InChI Key
HNNIWKQLJSNAEQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCOC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12
Assay (unspecified)
98.0% min.
Loss on Drying
0.5% max.
Melting Point
157-160?
pH
4.0-6.0
Sulfated Ash
0.1% max.

Description

Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Nonsteroidal analgesic and anti-inflammatory compound
  • A prostaglandin synthase inhibitor
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text