Glucose-6-phosphate dehydrogenase, Thermo Scientific Chemicals
Glucose-6-phosphate dehydrogenase, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glucose-6-phosphate dehydrogenase, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 kU
Packaging:
Plastic bottle
Catalog number J61181.4I
also known as J61181-4I
Price (USD)/ Each
85.65
Online exclusive
95.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
1 kU
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
85.65
Online exclusive
95.30 
Save 9.65 (10%)
Add to cart
Glucose-6-phosphate dehydrogenase, Thermo Scientific Chemicals
Catalog numberJ61181.4I
Price (USD)/ Each
85.65
Online exclusive
95.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS9001-40-5
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)% 6-Phosphogluconate dehydrogenase: 0.05% max.
Assay (unspecified)% Creatine phosphokinase: 0.05% max.
Assay (unspecified)% Hexokinase: 0.05% max.
Assay (unspecified)% Phosphoglucoisomerase: 0.05% max.
Unit definitionOne unit reduces one micromole of NADP per minute at 25?C, pH 8.0.
View more
Glucose-6-phosphate dehydrogenase, yeast converts p-glucose-6-phosphate to p-glucono-d-lactone-6-phosphate in presence of NADP+ or NAD+ for the determination of blood glucose or creatine kinase. It is used to investigate the effect of dehydroepiandrosterone on cell growth and mitochondrial function in TM-3 cells, in bovine follicular cells and in the incidence of apoptosis and transcript abundance. It is also useful to test ketose reductase activity in developing maize endosperm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only