Thermo Scientific Chemicals

Fluoxetine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J61197.MD
250 mg, Each
Thermo Scientific Chemicals

Fluoxetine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J61197.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61197.MD
also known as J61197-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56296-78-7
IUPAC Name
hydrogen methyl({3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl})amine chloride
Molecular Formula
C17H19ClF3NO
InChI Key
GIYXAJPCNFJEHY-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CNCCC(OC1=CC=C(C=C1)C(F)(F)F)C1=CC=CC=C1
Appearance (Color)
White
Assay from Supplier's CofA
≥98.5% (HPLC)
Form
Crystalline powder

Description

Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

Solubility
Soluble in dimethylsulfoxide and water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text