Thermo Scientific Chemicals

Ribonucleic acid from Baker's yeast, Thermo Scientific Chemicals

Catalog number: J61215.03
1 g, Each
Thermo Scientific Chemicals

Ribonucleic acid from Baker's yeast, Thermo Scientific Chemicals

Catalog number: J61215.03
1 g, Each
Quantity
Catalog number: J61215.03
also known as J61215-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63231-63-0
IUPAC Name
Ribonucleic acid
MDL Number
MFCD00132196
Appearance (Color)
Light cream to cream
Form
Powder
Assay (special)
% of native RNA: Report value
Assay (special)
Substrate suitability: satisfactory for Ribonuclease assay

Description

Ribonucleic acid (RNA) from baker?s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribonucleic acid (RNA) from baker′s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

Solubility
It is soluble in water.

Notes
Prepared by a modification of Crestfield, A.M., et al., J. Biol. Chem., 216, 185 (1955). Suitable for use as a ribonuclease substrate. Not suitable for use as a carrier in nucleic acid purification and precipitation
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text