Thermo Scientific Chemicals

Dextran, MW ca 20,000, Thermo Scientific Chemicals

Catalog number: J61216.36
500 g, Each
Thermo Scientific Chemicals

Dextran, MW ca 20,000, Thermo Scientific Chemicals

Catalog number: J61216.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61216.36
also known as J61216-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-54-0
IUPAC Name
Dextran
Molecular Formula
(C6H9O5)n(C6H10O5)m
SMILES
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
MDL Number
MFCD00130935
Form
White to off white powder
Assay (unspecified)
Specific rotation (2% in water) [|b]: NLT + 148?
Loss on Drying
≤ 7.0 %
pH
4.5 to 7.0 (10% solution)
Solution Test
1% in water

Description

Low molecular weight dextran is useful as a stabilizer, volume extender, lubricant, binding platform, and matrix component. Higher molecular weight dextrans have differing viscosities, aggregation and permeation properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text