Thermo Scientific Chemicals

(-)-Epicatechin, Thermo Scientific Chemicals

Catalog number: J61218.FPL
20 mg, Each
Thermo Scientific Chemicals

(-)-Epicatechin, Thermo Scientific Chemicals

Catalog number: J61218.FPL
20 mg, Each
Quantity
Catalog number: J61218.FPL
also known as J61218-F+
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
490-46-0
IUPAC Name
(2R,3R)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol
Molecular Formula
C15H14O6
InChI Key
PFTAWBLQPZVEMU-UKRRQHHQSA-N
SMILES
O[C@@H]1CC2=C(O)C=C(O)C=C2O[C@@H]1C1=CC=C(O)C(O)=C1
Assay (unspecified)
>95%

Description

An antioxidant and natural product from green tea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text