Thermo Scientific Chemicals

Concanavalin A, Thermo Scientific Chemicals

Catalog number: J61221.MC
100 mg, Each
Thermo Scientific Chemicals

Concanavalin A, Thermo Scientific Chemicals

Catalog number: J61221.MC
100 mg, Each
Quantity
Catalog number: J61221.MC
also known as J61221-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11028-71-0
Form
Salt free lyophilized powder
Appearance (Color)
White to off-white
Assay from Suppliers CofA
≥75%

Description

Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.

Solubility
Slightly soluble in water and phosfate buffered saline.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text