Thermo Scientific Chemicals

Tolfenamic acid, 99+%, Thermo Scientific Chemicals

Catalog number: J61256.09
10 g, Each
Thermo Scientific Chemicals

Tolfenamic acid, 99+%, Thermo Scientific Chemicals

Catalog number: J61256.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61256.09
also known as J61256-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13710-19-5
IUPAC Name
2-[(4-chloro-3-methylphenyl)amino]benzoic acid
Molecular Formula
C14H12ClNO2
InChI Key
QDNMBJXNLJFNHT-UHFFFAOYSA-N
SMILES
CC1=CC(NC2=CC=CC=C2C(O)=O)=CC=C1Cl
Thin Layer Chromatography
95.0% min.

Description

Tolfenamic acid is a non-steroidal anti-inflammatory agent. It interferes with synthesis of β-amyloid precursor protein, and thus Aβ peptides, by promoting degradation of an essential transcription factor. It inhibits fMLP- and A23187-induced Ca2+ influx in human PMNL with an IC50 value of approximately 20 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (slightly), acetone (∼10 mg/ml), DMSO (52 mg/ml at 25°C), methanol (∼10 mg/ml) and ethanol (50 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text