Kanamycin monosulfate
Kanamycin monosulfate
Kanamycin monosulfate
Kanamycin monosulfate
Thermo Scientific Chemicals

Kanamycin monosulfate

Kanamycin monosulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity. | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61272.09
also known as J61272-09
Price (USD)
118.65
Special offer
Online exclusive
Ends: 26-Sep-2025
146.00
Save 27.35 (19%)
Each
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
118.65
Special offer
Online exclusive
Ends: 26-Sep-2025
146.00
Save 27.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS25389-94-0
IUPAC Name(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-3-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}oxane-3,4,5-triol; sulfuric acid
Molecular FormulaC18H38N4O15S
InChI KeyOOYGSFOGFJDDHP-KMCOLRRFSA-N
SMILESOS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation+112.0 to +123° (c=1, water)
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Loss on Drying≤4.0%
View more
Ask our AI about this Product
Antibiotic. Inhibits translocation during protein biosynthesis.Kanamycin monosulfate acts as an antibiotic and used to treat various infections. It is involved in the translation of prokaryotic ribosomes arises due to bacterial cell death. It is used as a selective agent in molecular biology in order to isolate transformants that are expressing a kanamycin resistance gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Kanamycin monosulfate is the sulfate salt form of kanamycin A
  • It was isolated from Streptomyces kanamyceticus
  • This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

  • Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
  • This compound is also involved in translating prokaryotic ribosomes
  • In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated
RUO – Research Use Only

General References:

  1. Patel, K. N.; Limgavkar, R. S.; Raval, H. G.; Patel, K. G.; Gandhi, T. R. High-Performance Liquid Chromatographic Determination of Cefalexin Monohydrate and Kanamycin Monosulfate with Precolumn Derivatization. J. Liq. Chromatogr. Related Technol. 2015, 38 (6), 716-721.
  2. Chi, Z.; Rong, Y. J.; Li, Y.; Tang, M. J.; Chi, Z. M. Biosurfactins production by Bacillus amyloliquefaciens R3 and their antibacterial activity against multi-drug resistant pathogenic E. coli. Bioprocess. Biosyst. Eng. 2015, 38 (5), 853-861.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
T02L014Certificate of AnalysisJul 03, 2025J61272.14, J61272.09, J61272.22
Q08L023Certificate of AnalysisMay 17, 2025J61272.14, J61272.09, J61272.22
P06L011Certificate of AnalysisMar 10, 2025J61272.14, J61272.09, J61272.22
X24K031Certificate of AnalysisNov 11, 2024J61272.14, J61272.09, J61272.22
W10K010Certificate of AnalysisSep 24, 2024J61272.14, J61272.09, J61272.22
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection