Paromomycin sulfate, Thermo Scientific Chemicals
Paromomycin sulfate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Paromomycin sulfate, Thermo Scientific Chemicals

Inhibits initiation and elongation during protein synthesis
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61274.03
also known as J61274-03
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.40 
Save 7.75 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.40 
Save 7.75 (15%)
Add to cart
Paromomycin sulfate, Thermo Scientific Chemicals
Catalog numberJ61274.03
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.40 
Save 7.75 (15%)
-
Add to cart
Chemical Identifiers
CAS1263-89-4
IUPAC Name(2S,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(2R,3S,4R)-5-{[(1R,3S,5R,6S)-3,5-diamino-2-{[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-hydroxycyclohexyl]oxy}-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Molecular FormulaC23H47N5O18S
InChI KeyLJRDOKAZOAKLDU-UMIPPVEZSA-N
SMILESOS(O)(=O)=O.NC[C@@H]1O[C@H](O[C@@H]2[C@@H](CO)OC(O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)C3O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)[C@H](N)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
FormPowder
Assay from Supplier's CofA≥675 U/mg
Loss on Drying≤5.0%
Paromomycin sulfate salt has been used as a reference compound in antileishmanial activity, RNA-binding ligand and interacts with aptamer. This interaction prevents the binding (and cutting) of dicer to RNA duplex. It has also been used in cell culture applications at 100mg/L. Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Paromomycin sulfate salt has been used as a reference compound in antileishmanial activity, RNA-binding ligand and interacts with aptamer. This interaction prevents the binding (and cutting) of dicer to RNA duplex. It has also been used in cell culture applications at 100mg/L. Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Begley U et al. A human tRNA methyltransferase 9-like protein prevents tumour growth by regulating LIN9 and HIF1-a. EMBO Mol Med . 2013, 5,(3), 366-83.
  2. Goossens J et al. Porcine intestinal epithelial barrier disruption by the Fusarium mycotoxins deoxynivalenol and T-2 toxin promotes transepithelial passage of doxycycline and paromomycin. BMC Vet Res. 2012, 8,(245), 6148-8.