Thermo Scientific Chemicals

Paromomycin sulfate, Thermo Scientific Chemicals

Catalog number: J61274.06
5 g, Each
Thermo Scientific Chemicals

Paromomycin sulfate, Thermo Scientific Chemicals

Catalog number: J61274.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61274.06
also known as J61274-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1263-89-4
IUPAC Name
(2S,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(2R,3S,4R)-5-{[(1R,3S,5R,6S)-3,5-diamino-2-{[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-hydroxycyclohexyl]oxy}-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Molecular Formula
C23H47N5O18S
InChI Key
LJRDOKAZOAKLDU-UMIPPVEZSA-N
SMILES
OS(O)(=O)=O.NC[C@@H]1O[C@H](O[C@@H]2[C@@H](CO)OC(O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)C3O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)[C@H](N)[C@@H](O)[C@@H]1O
Form
Powder
Appearance (Color)
White to pale cream to pale yellow
Assay from Supplier's CofA
≥675 U/mg
Loss on Drying
≤5.0%

Description

Paromomycin sulfate salt has been used as a reference compound in antileishmanial activity, RNA-binding ligand and interacts with aptamer. This interaction prevents the binding (and cutting) of dicer to RNA duplex. It has also been used in cell culture applications at 100mg/L. Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Paromomycin sulfate salt has been used as a reference compound in antileishmanial activity, RNA-binding ligand and interacts with aptamer. This interaction prevents the binding (and cutting) of dicer to RNA duplex. It has also been used in cell culture applications at 100mg/L. Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text