Thermo Scientific Chemicals

(+/-)-Octopamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J61281.06
5 g, Each
Thermo Scientific Chemicals

(+/-)-Octopamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J61281.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61281.06
also known as J61281-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
770-05-8
IUPAC Name
4-(2-amino-1-hydroxyethyl)phenol hydrochloride
Molecular Formula
C8H12ClNO2
InChI Key
PUMZXCBVHLCWQG-UHFFFAOYNA-N
SMILES
Cl.NCC(O)C1=CC=C(O)C=C1
Assay (unspecified)
98.5% min.

Description

Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

Solubility
It is soluble in water.

Notes
Handle under argon. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text