Thermo Scientific Chemicals

Amoxicillin trihydrate, Thermo Scientific Chemicals

Catalog number: J61290.14
25 g, Each
Thermo Scientific Chemicals

Amoxicillin trihydrate, Thermo Scientific Chemicals

Catalog number: J61290.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61290.14
also known as J61290-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61336-70-7
IUPAC Name
(2S,5R,6R)-6-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
Molecular Formula
C16H25N3O8S
InChI Key
MQXQVCLAUDMCEF-CWLIKTDRSA-N
SMILES
O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=C(O)C=C3)C(=O)N2[C@H]1C(O)=O
Impurity content
≤5.0 %
Water Content (Karl Fischer Titration)
11.5-14.5%
pH
3.5-6.0
Appearance (Color)
White to pale cream
Form
Powder or crystals or crystalline powder

Description

Amoxicillin trihydratesemi-synthetic antibiotic related to Penicillin which has antibacterial poperties. It is used for treatment of a variety of bacterial infections. It is used as a pharmaceutical product as a antibiotic agent, anti-infective drug raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amoxicillin trihydratesemi-synthetic antibiotic related to Penicillin which has antibacterial poperties. It is used for treatment of a variety of bacterial infections. It is used as a pharmaceutical product as a antibiotic agent, anti-infective drug raw materials.

Solubility
Soluble in water at 3.4mg/ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text