LB Agar plate with 50μg/mL Ampicillin
This product is animal origin and can only be shipped within the United States
Product Image
Thermo Scientific Chemicals
LB Agar plate with 50μg/mL Ampicillin
Have Questions?
Quantity:
20 Plates
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61314.EQF
also known as J61314-=F
Price (USD)/ Each
-
Quantity:
20 Plates
Request bulk or custom format
Specifications
Recommended StorageKeep cold
FormSolid
Media TypeLB Agar
For Use With (Application)For growth of Escherichia coli, plasmid and other bacterial cultures
Packaging TypePlate
View more
Miller’s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. It is used with recombinant strains of E.coli containing plasmids with the resistance gene to Ampicillin (bla or ampR) such as pBluescript, pGEM, or the pUC series of plasmids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used for growth of Escherichia coli, plasmid and other bacterial cultures
RUO – Research Use Only
Contents & Storage
2°C to 8°C