Carboxy-PTIO potassium salt, 98+%, Thermo Scientific Chemicals
Carboxy-PTIO potassium salt, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carboxy-PTIO potassium salt, 98+%, Thermo Scientific Chemicals

Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Catalog number J61316.MF
also known as J61316-MF
Price (USD)/ Each
397.65
Online exclusive
442.00 
Save 44.35 (10%)
-
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)/ Each
397.65
Online exclusive
442.00 
Save 44.35 (10%)
Add to cart
Carboxy-PTIO potassium salt, 98+%, Thermo Scientific Chemicals
Catalog numberJ61316.MF
Price (USD)/ Each
397.65
Online exclusive
442.00 
Save 44.35 (10%)
-
Add to cart
Chemical Identifiers
CAS148819-94-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.0% min.
Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only