Ciprofloxacin, 98%, Thermo Scientific Chemicals
Ciprofloxacin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ciprofloxacin, 98%, Thermo Scientific Chemicals

An anti-bacterial agent effective against anaerobic bacteria
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61317.06
also known as J61317-06
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.40 
Save 7.75 (16%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.40 
Save 7.75 (16%)
Add to cart
Ciprofloxacin, 98%, Thermo Scientific Chemicals
Catalog numberJ61317.06
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.40 
Save 7.75 (16%)
-
Add to cart
Chemical Identifiers
CAS85721-33-1
IUPAC Name1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
Molecular FormulaC17H18FN3O3
InChI KeyMYSWGUAQZAJSOK-UHFFFAOYSA-N
SMILESOC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤1.0%
Identification (FTIR)Conforms
FormPowder or crystalline powder
Appearance (Color)White to pale cream to pale yellow
Assay from Supplier's CofA≥97.5% (HPLC)
Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.

Solubility
Soluble in 0.1N HCl at 25mg/ml. Poorly soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

General References:

  1. Nix DE.; DeVito JM. Ciprofloxacin and norfloxacin, two fluoroquinolone antimicrobials. Clin Pharm. 1987, 6, (2), 105-17.
  2. Sen, et al. Anxiogenic potential of ciprofloxacin and norfloxacin in rats. Singapore Med. J. 2007, 48, (11), 1028-30.