Thermo Scientific Chemicals

Ciprofloxacin, 98%, Thermo Scientific Chemicals

Catalog number: J61317.14
25 g, Each
Thermo Scientific Chemicals

Ciprofloxacin, 98%, Thermo Scientific Chemicals

Catalog number: J61317.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61317.14
also known as J61317-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85721-33-1
IUPAC Name
1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
Molecular Formula
C17H18FN3O3
InChI Key
MYSWGUAQZAJSOK-UHFFFAOYSA-N
SMILES
OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Loss on Drying
≤1.0%
Identification (FTIR)
Conforms
Form
Powder or crystalline powder
Appearance (Color)
White to pale cream to pale yellow
Assay from Supplier's CofA
≥97.5% (HPLC)

Description

Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.

Solubility
Soluble in 0.1N HCl at 25mg/ml. Poorly soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text