Thermo Scientific Chemicals

Dobutamine hydrochloride, Thermo Scientific Chemicals

Catalog number: J61333.03
1 g, Each
Thermo Scientific Chemicals

Dobutamine hydrochloride, Thermo Scientific Chemicals

Catalog number: J61333.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61333.03
also known as J61333-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
49745-95-1
Form
Powder
Appearance (Color)
White to off-white
Assay (HPLC)
≥98.0%

Description

Dobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1M.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1 μM.

Solubility
Soluble in water or ethanol with warming. Also soluble in methanol

Notes
Solutions oxidize readily at pH greater than 11. Protect solutions from light
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text