Thermo Scientific Chemicals

4-Fluoro-7-nitrobenzofurazan, Thermo Scientific Chemicals

Catalog number: J61336.MC
100 mg, Each
Thermo Scientific Chemicals

4-Fluoro-7-nitrobenzofurazan, Thermo Scientific Chemicals

Catalog number: J61336.MC
100 mg, Each
Quantity
Catalog number: J61336.MC
also known as J61336-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29270-56-2
IUPAC Name
4-fluoro-7-nitro-2,1,3-benzoxadiazole
Molecular Formula
C6H2FN3O3
InChI Key
PGZIDERTDJHJFY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(F)C2=NON=C12
Appearance (Color)
White to yellow or green
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥99.0% (HPLC)

Description

4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the following studies, Synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, Synthesis of functionalized hydroxynaphthofurazan, Spectrophotometric and spectrofluorometric determination of clemastine hydrogen fumarate, loratadine, losartan potassium and ramipril in pharmaceutical formulations. Synthesis of 7-nitrobenzofurazan (NBD)-labeled maleimide, via Diels-Alder reaction. NBD-Cl is widely used to label peptides, proteins, drugs and other biomolecules. It is also a popular derivatizing reagent for HPLC analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the following studies, Synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, Synthesis of functionalized hydroxynaphthofurazan, Spectrophoto
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text