Thermo Scientific Chemicals

Laemmli SDS sample buffer, reducing (6X)

Catalog number: J61337.AD
50 mL, Each
Thermo Scientific Chemicals

Laemmli SDS sample buffer, reducing (6X)

Catalog number: J61337.AD
50 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61337.AD
also known as J61337-AD
Price (USD)
Quantity
-
Have Questions?
Composition
375mM Tris-HCl/Tris Base, 9% SDS (w/v), 50% Glycerol (v/v), 9% 2-mercaptoethanol (v/v), 0.075% Bromophenol blue (w/v)
Appearance (Color)
Clear blue
Form
Liquid
pH
6.6-7.0 (1X)

Description

For protein sample preparation to be used in the Laemmli SDS-PAGE system. Laemmli SDS sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Further, it is used for polyacrylamide protein gel analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • This compound is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 9% β-mercaptoethanol, 0.03% bromophenol blue
  • The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds

Application

  • Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
  • This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
  • It is also used for polyacrylamide protein gel analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text