Paxilline, 97+%, Thermo Scientific Chemicals
Paxilline, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Paxilline, 97+%, Thermo Scientific Chemicals

Potent blocker of high-conductance calcium-activated potassium (BKCa) channels
Have Questions?
Quantity:
5 mg
Catalog number J61341.LB0
also known as J61341-#0
Price (USD)/ Each
250.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
250.00
Add to cart
Paxilline, 97+%, Thermo Scientific Chemicals
Catalog numberJ61341.LB0
Price (USD)/ Each
250.00
-
Add to cart
Specifications
Chemical Name or MaterialPaxilline
CAS57186-25-1
Health Hazard 1H301+H311-H315-H318-H330-H335
Health Hazard 2GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
View more
Potent blocker of high-conductance calcium-activated potassium (BKCa) channels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only