Azure A, Thermo Scientific Chemicals
Azure A, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azure A, Thermo Scientific Chemicals

For staining cell granules and tissues
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number J61346.22
also known as J61346-22
Price (USD)/ Each
75.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
75.00
Add to cart
Azure A, Thermo Scientific Chemicals
Catalog numberJ61346.22
Price (USD)/ Each
75.00
-
Add to cart
Chemical Identifiers
CAS531-53-3
IUPAC Name3-amino-7-(dimethylamino)-5λ⁴-phenothiazin-5-ylium chloride
Molecular FormulaC14H14ClN3S
InChI KeyPGWTYMLATMNCCZ-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=CC(N)=CC3=[S+]C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Absorptivity:>1120, 1%/1cm at lambda max 630nm
Assay (unspecified)Dye Content: 70-80%
Loss on Drying<8.0%
For staining cell granules and tissues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only