TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.
Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
NE Good.; GD Winget.; W Winter.; TN Connolly. Hydrogen ion buffers for biological research. Biochemistry. 1966, 5, (2), 467-477.
S Matsushita. Coloring conditions of thiobarbituric acid test for detecting lipid hydro peroxides. Lipids. 1980, 15, (3), 137-140.