N-Methyl-D-aspartic acid, 98+%, Thermo Scientific Chemicals
N-Methyl-D-aspartic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyl-D-aspartic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
50 mg
Catalog number J61361.MC
also known as J61361-MC
Price (USD)/ Each
133.00
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
133.00
Add to cart
N-Methyl-D-aspartic acid, 98+%, Thermo Scientific Chemicals
Catalog numberJ61361.MC
Price (USD)/ Each
133.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Methyl-D-aspartic acid
Melting Point187°C to 192°C
CAS6384-92-5
ColorWhite
Recommended StorageKeep cold
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only