Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific™

SM buffer, pH 7.5

Catalog number: J61367.AE
100 mL, Each
Thermo Scientific™

SM buffer, pH 7.5

Catalog number: J61367.AE
100 mL, Each
Quantity
Catalog number: J61367.AE
also known as J61367-AE
Price (USD)
Have Questions?
Appearance (Color)
Clear colorless
Form
Liquid
Concentration
Geletin: 0.02%
Concentration
Magnesium Sulfate Heptahydrate: 8 mM
pH
7.3-7.7

Description

SM buffer is mainly used in molecular biology lab. It is used for routine manipulation of phage suspensions. The gelatin in SM buffer stabilizes lambda phage particles during storage. is mainly used in molecular biology lab.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
SM buffer is mainly used in molecular biology lab. It is used for routine manipulation of phage suspensions. The gelatin in SM buffer stabilizes lambda phage particles during storage. is mainly used in molecular biology lab.

Solubility
Not miscible in water.

Notes
50mM Tris-HCl, 8mM magnesium sulfate, 100mM sodium chloride, and 0.01% gelatin, pH7.5
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text