Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals
Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals

Inhibitor of thrombaxane synthetase
Have Questions?
Quantity:
5 mg
Catalog number J61378.LB0
also known as J61378-#0
Price (USD)/ Each
118.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
118.00
Add to cart
Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals
Catalog numberJ61378.LB0
Price (USD)/ Each
118.00
-
Add to cart
Specifications
Quantity5 mg
DescriptionWhite to Off-white
For Use With (Application)Inhibitor of thrombaxane synthetase
Recommended StorageStore at -20°C
Assay≥99%
View more
Inhibitor of thrombaxane synthetase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C