Chemical Identifiers
CAS2181-04-6
IUPAC Namepotassium 3-[(1R,3aS,3bR,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]propanoate
Molecular FormulaC22H29KO4
InChI KeyJTZQCHFUGHIPDF-RYVBEKKQSA-M
SMILES[K+].C[C@]12CC[C@H]3[C@@H](C=CC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@]2(O)CCC([O-])=O
View more