Thermo Scientific Chemicals

Alendronate sodium trihydrate, 97%, Thermo Scientific Chemicals

Catalog number: J61397.06
5 g, Each
Thermo Scientific Chemicals

Alendronate sodium trihydrate, 97%, Thermo Scientific Chemicals

Catalog number: J61397.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61397.06
also known as J61397-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121268-17-5
IUPAC Name
sodium trihydrate hydrogen (4-amino-1-hydroxy-1-phosphonobutyl)phosphonate
Molecular Formula
C4H18NNaO10P2
InChI Key
DCSBSVSZJRSITC-UHFFFAOYNA-M
SMILES
O.O.O.[Na+].NCCCC(O)(P(O)(O)=O)P(O)([O-])=O
Form
Powder or crystals or crystalline powder
Identification (FTIR)
Conforms
Assay from Supplier's CofA
≥96.0%
Appearance (Color)
White to pale cream
Loss on Drying
14.0-19.0%

Description

Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.

Solubility
Soluble in water at 10mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text