Thermo Scientific Chemicals

Leu-Enkephalin

Catalog number: J61408.MB
25mg, Each
Thermo Scientific Chemicals

Leu-Enkephalin

Catalog number: J61408.MB
25mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61408.MB
also known as J61408-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58822-25-6
Form
Lyophilized powder
Assay from Supplier's CofA
≥94.0%
Molecular Weight
Consistent with structure (theoretical 555.62 g/mol)
Appearance (Color)
White

Description

Endogenous opioid neurotransmitter/neuromodulator that is an agonist at ? and δ opioid receptors, localization in brain parallels the localization of δ receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Endogenous opioid neurotransmittereuromodulator that is an agonist at μ and δ opioid receptors, localization in brain parallels the localization of δ receptors.

Solubility
Soluble in water (1 mg/ml ).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text