Thermo Scientific Chemicals

Clindamycin hydrochloride monohydrate, Thermo Scientific Chemicals

Catalog number: J61409.06
5 g, Each
Thermo Scientific Chemicals

Clindamycin hydrochloride monohydrate, Thermo Scientific Chemicals

Catalog number: J61409.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61409.06
also known as J61409-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58207-19-5
IUPAC Name
N-{2-chloro-1-[3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl}-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride
Molecular Formula
C18H34Cl2N2O5S
InChI Key
AUODDLQVRAJAJM-UHFFFAOYNA-N
SMILES
Cl.CCCC1CC(N(C)C1)C(=O)NC(C(C)Cl)C1OC(SC)C(O)C(O)C1O
Appearance (Color)
White to cream
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥98.0%

Description

Clindamycin hydrochloride monohydrate is an antibacterial agent that is a semisynthetic analog of lincomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Clindamycin is a lincosamide antibiotic that disrupts protein synthesis by interfering with the aminoacyl-tRNA ribosome complex

• Clindamycin decreases sebaceous gland activity and blocks the neuromuscular junction

Application

• Clindamycin is commonly used in laboratory applications to inhibit the growth anaerobic microbes, but has some effectiveness against aerobic microbes as well

• It can be used in the laboratory study of cell culture for resistance testing, or in antibiotic allergic reaction studies

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text