2'-Deoxyinosine, Thermo Scientific Chemicals
2'-Deoxyinosine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Deoxyinosine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Catalog number J61411.14
also known as J61411-14
Price (USD)/ Each
380.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
380.00
Add to cart
2'-Deoxyinosine, Thermo Scientific Chemicals
Catalog numberJ61411.14
Price (USD)/ Each
380.00
-
Add to cart
Specifications
Chemical Name or Material2'-Deoxyinosine
CAS890-38-0
Melting Point>250°C
ColorWhite
Recommended StorageKeep cold
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only